5-Amino-1H-tetrazole ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A0427-25G A0427-500G A0427-100G
TCIAA0427-25GEA 50.6 EUR
TCIAA0427-25G TCIAA0427-500G TCIAA0427-100G
5-Amino-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-Amino-1H-tetrazole
Formule: CH₃N₅
Kookpunt: 387 °C (1013 hPa)
Smeltpunt: 216…217 °C
Dichtheid: 1,51 g/cm³ (20 °C)
MDL Nummer: MFCD00005248
CAS nummer: 4418-61-5
EINECS: 224-581-7
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c123311389fe180c83b9808dc4670a7ae5ed9d2&vid=3c123311389fe180c83b9808dc4670a7ae5ed9d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR