N-Acetyl-S-benzyl-DL-cysteine ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A0730-1G
TCIAA0730-1GEA 326 EUR
TCIAA0730-1G
N-Acetyl-S-benzyl-DL-cysteine ≥98.0% (by titrimetric analysis)
N-Acetyl-S-benzyl-DL-cysteine
Formule: C₁₂H₁₅NO₃S
MDL Nummer: MFCD00067460
CAS nummer: 19538-71-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 155.0 to 158.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR