Ethyl DL-3-acetoxybutyrate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A0831-25ML A0831-5ML
TCIAA0831-25MLEA 105 EUR
TCIAA0831-25ML TCIAA0831-5ML
Ethyl DL-3-acetoxybutyrate ≥98.0% (by GC)
Ethyl DL-3-acetoxybutyrate
Formule: C₈H₁₄O₄
Kookpunt: 94°C
MDL Nummer: MFCD00059378
CAS nummer: 27846-49-7
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR