2-Acetyl-1H-pyrrole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A0894-25G A0894-5G
TCIAA0894-25GEA 92.4 EUR
TCIAA0894-25G TCIAA0894-5G
2-Acetyl-1H-pyrrole ≥98.0% (by GC)
2-Acetyl-1H-pyrrole
Formule: C₆H₇NO
Kookpunt: 220°C
Smeltpunt: 63…65 °C
MDL Nummer: MFCD00005220
CAS nummer: 1072-83-9
EINECS: 214-016-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8e45af97d63d1cf2199f83d7a459b27e2d0d974&vid=c8e45af97d63d1cf2199f83d7a459b27e2d0d974&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 89.0 to 93.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR