5-Phenyl-o-anisidine ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A0958-25G A0958-5G
TCIAA0958-25GEA 289 EUR
TCIAA0958-25G TCIAA0958-5G
5-Phenyl-o-anisidine ≥98.0% (by titrimetric analysis)
5-Phenyl-o-anisidine
Formule: C₁₃H₁₃NO
MDL Nummer: MFCD00007790
CAS nummer: 39811-17-1
EINECS: 254-639-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7720f1e9770633c6cc3e7c2d26965b01ea1ca015&vid=7720f1e9770633c6cc3e7c2d26965b01ea1ca015&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light red to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 83.0 to 86.0 deg-C
Solubility in Methanol almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR