1,6-Anhydro-β-D-glucose ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A1074-1G A1074-5G
TCIAA1074-1GEA 29.7 EUR
TCIAA1074-1G TCIAA1074-5G
1,6-Anhydro-β-D-glucose ≥99.0% (by GC)
1,6-Anhydro-beta-D-glucose
Formule: C₆H₁₀O₅
MDL Nummer: MFCD00063248
CAS nummer: 498-07-7
EINECS: 207-855-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 177.0 to 184.0 deg-C
Specific rotation [a]20/D -65.0 to -68.0 deg (C=2, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR