(R)-(+)-3-Aminopyrrolidine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A1167-25G A1167-5G A1167-1G
TCIAA1167-25GEA 831 EUR
TCIAA1167-25G TCIAA1167-5G TCIAA1167-1G
(R)-(+)-3-Aminopyrrolidine ≥98.0% (by GC, titration analysis)
(R)-(+)-3-Aminopyrrolidine
Formule: C₄H₁₀N₂
Vlampunt: 64°C
MDL Nummer: MFCD00143190
CAS nummer: 116183-82-5
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=555dc9d053cf176f12cfae06a11c171717b37833&vid=555dc9d053cf176f12cfae06a11c171717b37833&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 99.0 ee%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR