2'-Amino-4',5'-methylenedioxyacetophenone ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A1356-5G
TCIAA1356-5GEA 51.7 EUR
TCIAA1356-5G
2'-Amino-4',5'-methylenedioxyacetophenone ≥98.0% (by GC, titration analysis)
2'-Amino-4',5'-methylenedioxyacetophenone
Formule: C₉H₉NO₃
MDL Nummer: MFCD00075855
CAS nummer: 28657-75-2
EINECS: 249-131-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd1d82fcd3d047ae4548620dc710ffefa7445799&vid=cd1d82fcd3d047ae4548620dc710ffefa7445799&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR