N-Boc-1,3-propanediamine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A1372-1G A1372-5G
TCIAA1372-1GEA 35.2 EUR
TCIAA1372-1G TCIAA1372-5G
N-Boc-1,3-propanediamine ≥98.0% (by GC)
N-Boc-1,3-propanediamine
Formule: C₈H₁₈N₂O₂
MDL Nummer: MFCD00210021
CAS nummer: 75178-96-0
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR