N-Acetyl-2-(4-nitrophenyl)ethylamine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A1494-25G A1494-5G
TCIAA1494-25GEA 362 EUR
TCIAA1494-25G TCIAA1494-5G
N-Acetyl-2-(4-nitrophenyl)ethylamine ≥98.0% (by GC)
N-Acetyl-2-(4-nitrophenyl)ethylamine
Formule: C₁₀H₁₂N₂O₃
MDL Nummer: MFCD00024805
CAS nummer: 6270-07-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 142.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR