3'-Aminoacetanilide ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: N-(3-Aminophenyl)acetamide, 3-Acetamidoaniline, N-Acetyl-1,3-phenylenediamine, m-Aminoacetanilide

A1533-25G
TCIAA1533-25GEA 35.2 EUR
TCIAA1533-25G
3'-Aminoacetanilide ≥98.0% (by HPLC, titration analysis)
3'-Aminoacetanilide
Formule: C₈H₁₀N₂O
MW: 150.18 g/mol
Kookpunt: 388
MDL Nummer: MFCD00025229
CAS nummer: 102-28-3
EINECS: 203-021-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53292f8b07260cd89da25ea69bca3894f4bcf101&vid=53292f8b07260cd89da25ea69bca3894f4bcf101&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR