6-Aminoindole ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A2067-1G
TCIAA2067-1GEA 115 EUR
TCIAA2067-1G
6-Aminoindole ≥98.0% (by GC, titration analysis)
6-Aminoindole
Formule: C₈H₈N₂
Kookpunt: 166°C
Smeltpunt: 65…66 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02093959
CAS nummer: 5318-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2d8195d3b01903b9488988b40c1c144b49b8ba1&vid=c2d8195d3b01903b9488988b40c1c144b49b8ba1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR