2-Amino-3-chloropyrazine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 3-Chloropyrazin-2-amine, 3-Chloro-2-pyrazinamine

A2143-1G A2143-5G
TCIAA2143-1GEA 29.7 EUR
TCIAA2143-1G TCIAA2143-5G
2-Amino-3-chloropyrazine ≥98.0% (by GC)
2-Amino-3-chloropyrazine
Formule: C₄H₄ClN₃
MW: 129.55 g/mol
MDL Nummer: MFCD04114305
CAS nummer: 6863-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63963ec1a076a133f85426a90bc7a4393ee5428c&vid=63963ec1a076a133f85426a90bc7a4393ee5428c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR