2-Amino-6-chloropyrazine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A2145-1G A2145-5G
TCIAA2145-1GEA 25.3 EUR
TCIAA2145-1G TCIAA2145-5G
2-Amino-6-chloropyrazine ≥98.0% (by GC)
2-Amino-6-chloropyrazine
Formule: C₄H₄ClN₃
MW: 129.55 g/mol
MDL Nummer: MFCD00055024
CAS nummer: 33332-28-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8945977bda314c011fa1657d8e29cae99a39181a&vid=8945977bda314c011fa1657d8e29cae99a39181a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 156.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR