6-Aminopicolinic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A2188-1G A2188-5G
TCIAA2188-1GEA 38.5 EUR
TCIAA2188-1G TCIAA2188-5G
6-Aminopicolinic acid ≥98.0% (by HPLC, titration analysis)
6-Aminopicolinic acid
Formule: C₆H₆N₂O₂
Smeltpunt: 315°C
MDL Nummer: MFCD00233711
CAS nummer: 23628-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3757533f75846ac5437fe52888ea482c9299bb22&vid=3757533f75846ac5437fe52888ea482c9299bb22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR