1-Allylimidazole ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A2207-25G
TCIAA2207-25GEA 56.1 EUR
TCIAA2207-25G
1-Allylimidazole ≥97.0% (by GC, titration analysis)
1-Allylimidazole
Formule: C₆H₈N₂
Kookpunt: 116°C
Vlampunt: 101°C
MDL Nummer: MFCD00037894
CAS nummer: 31410-01-2
EINECS: 250-618-1
UN: 3267
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69ce4ba6b7e4e1b5bf3cb7c4a752953251b342f8&vid=69ce4ba6b7e4e1b5bf3cb7c4a752953251b342f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR