Ammonium thioglycolate ∼60% in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Ammonium thioglycolate solution, Thioglycolic Acid Ammonium Salt, Ammonium Mercaptoacetate

A2217-25G A2217-500G
TCIAA2217-25GEA 17.6 EUR
TCIAA2217-25G TCIAA2217-500G
Ammonium thioglycolate ∼60% in water
Ammonium thioglycolate
Formule: C₂H₇NO₂S
MDL Nummer: MFCD00137451
CAS nummer: 5421-46-5
EINECS: 226-540-9
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (Iodometric titration) 57.0 to 63.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR