6-Aminocaproic acid ≥98.0% (by titrimetric analysis) for biochemical research

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid

A2255-25G A2255-5G
TCIAA2255-25GEA 28.6 EUR
TCIAA2255-25G TCIAA2255-5G
6-Aminocaproic acid ≥98.0% (by titrimetric analysis) for biochemical research
6-Aminocaproic acid
Formule: C₆H₁₃NO₂
MW: 131.17 g/mol
Kookpunt: 257
Smeltpunt: 204°C
Vlampunt: 207
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008238
CAS nummer: 60-32-2
EINECS: 200-469-3
Merck Index: 14,00432
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2a793eb45f7423924f34cd9682b528ab8bfa6e6&vid=63ddedc2e1806ac8ee8350ff5a503d5eced62a3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency
Blotting test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR