4',6-diamidino-2fenylindol dihydrocloride (DAPI) ≥90.0% (by HPLC) for biochemical research

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A2412-5MG
TCIAA2412-5MGEA 48.4 EUR
TCIAA2412-5MG
4',6-diamidino-2fenylindol dihydrocloride (DAPI) ≥90.0% (by HPLC) for biochemical research
4',6-diamidino-2fenylindol dihydrocloride (DAPI)
Formule: C₁₆H₁₅N₅·2HCl
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00012681
CAS nummer: 28718-90-3
EINECS: 249-186-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dd970dbedf5b446c85eabd71be8cf76412dbda7&vid=1a50cbeef1fdd1280d49ae3facb5454540e33233&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 90.0 area%
DNA staining test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR