S-(Acetamidomethyl)-N-(tert-butoxycarbonyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A2553-5G
TCIAA2553-5GEA 75.9 EUR
TCIAA2553-5G
S-(Acetamidomethyl)-N-(tert-butoxycarbonyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)
S-(Acetamidomethyl)-N-(tert-butoxycarbonyl)-L-cysteine
Formule: C₁₁H₂₀N₂O₅S
MDL Nummer: MFCD00038252
CAS nummer: 19746-37-3
EINECS: 243-267-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -38.0 to -41.0 deg (C=1、H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR