Fmoc-Glu(OAll)-OH ≥96.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A2895-1G A2895-5G
TCIAA2895-1GEA 46.2 EUR
TCIAA2895-1G TCIAA2895-5G
Fmoc-Glu(OAll)-OH ≥96.0% (by HPLC, titration analysis)
Fmoc-Glu(OAll)-OH
Formule: C₂₃H₂₃NO₆
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00190879
CAS nummer: 133464-46-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Melting point 125.0 to 129.0 deg-C
Specific rotation [a]20/D -15.0 to -19.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR