2-Acetyl-7-bromofluorene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A3061-1G
TCIAA3061-1GEA 105 EUR
TCIAA3061-1G
2-Acetyl-7-bromofluorene ≥98.0% (by GC)
2-Acetyl-7-bromofluorene
Formule: C₁₅H₁₁BrO
CAS nummer: 34172-50-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR