5-Amino-2,2-difluoro-1,3-benzodioxole ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A3107-1G A3107-5G
TCIAA3107-1GEA 51.7 EUR
TCIAA3107-1G TCIAA3107-5G
5-Amino-2,2-difluoro-1,3-benzodioxole ≥98.0% (by GC, titration analysis)
5-Amino-2,2-difluoro-1,3-benzodioxole
Formule: C₇H₅F₂NO₂
Kookpunt: 97°C
MDL Nummer: MFCD00190144
CAS nummer: 1544-85-0

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR