Sulphonazo III sodium salt ≥90.0% (by HPLC) for spectrophotometry

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A5010-1G A5010-5G
TCIAA5010-1GEA 79.2 EUR
TCIAA5010-1G TCIAA5010-5G
Sulphonazo III sodium salt ≥90.0% (by HPLC) for spectrophotometry
Sulphonazo III sodium salt

Spectrophotometric reagent for alkaline earth metals and indicator for the precipitation titration of SO4 with Ba

Formule: C₂₂H₁₆N₄O₁₄S₄
MDL Nummer: MFCD00003942
CAS nummer: 68504-35-8
EINECS: 270-924-9

Order Now

Specification Test Results

Appearance Dark green to Black powder to crystal
Purity (HPLC) min. 90.0 area%
Sensitiveness Colorimetric Test : Turn Purple with Barium

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR