Methylene blue 98,0 Titrimetric Ion association reagent for spectro anal

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

A5105-1G
TCIAA5105-1GEA 26.4 EUR
TCIAA5105-1G
Methylene blue 98,0 Titrimetric Ion association reagent for spectro anal
Methylene blue
Formule: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Smeltpunt: 180
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012111
CAS nummer: 61-73-4
EINECS: 200-515-2
Merck Index: 14,06060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3727a13bc2e773cc967b924077d93f420e2c8b8b&vid=2f056ffb64fbdfda736b654daba6a021ea7fc7cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Green to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Lambda max. 661.0 to 665.0 nm (EtOH (50%))
Absorbance min. 0.540 (2mg/L, EtOH (50%), 661.0 to 665.0 nm)
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR