2-Bromoethyl benzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Benzoic Acid 2-Bromoethyl Ester

B0065-25ML
TCIAB0065-25MLEA 272 EUR
TCIAB0065-25ML
2-Bromoethyl benzoate ≥98.0% (by GC)
2-Bromoethyl benzoate
Formule: C₉H₉BrO₂
MW: 229.07 g/mol
Kookpunt: 285°C
MDL Nummer: MFCD00059462
CAS nummer: 939-54-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR