4,4'-Bis(dimethylamino)diphenylmethane ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B0483-25G B0483-500G
TCIAB0483-25GEA 18.7 EUR
TCIAB0483-25G TCIAB0483-500G
4,4'-Bis(dimethylamino)diphenylmethane ≥98.0% (by GC, titration analysis)
4,4'-Bis(dimethylamino)diphenylmethane
Formule: C₁₇H₂₂N₂
MDL Nummer: MFCD00008317
CAS nummer: 101-61-1
EINECS: 202-959-2
Merck Index: 14,06176
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=728198952fbb9496c54726e0a2c491ce07c118de&vid=728198952fbb9496c54726e0a2c491ce07c118de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR