Methyl bromoacetate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B0533-25G B0533-500G B0533-100G
TCIAB0533-25GEA 17.6 EUR
TCIAB0533-25G TCIAB0533-500G TCIAB0533-100G
Methyl bromoacetate ≥98.0% (by GC)
Methyl bromoacetate
Formule: BrCH₂COOCH₃
Kookpunt: 144°C
Smeltpunt: –50 °C
Dichtheid: 1,635 g/cm³ (20 °C)
Vlampunt: 63°C
MDL Nummer: MFCD00000189
CAS nummer: 96-32-2
EINECS: 202-499-2
UN: 2643
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdf234cb7f0f8036e5b29c7a0eb8f33a1b674b6e&vid=bdf234cb7f0f8036e5b29c7a0eb8f33a1b674b6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR