1-Bromobutane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B0560-25G B0560-500G
TCIAB0560-25GEA 19.8 EUR
TCIAB0560-25G TCIAB0560-500G
1-Bromobutane ≥98.0% (by GC)
1-Bromobutane
Formule: BrH₂C(CH₂)₂CH₃
Kookpunt: 101°C
Smeltpunt: -112°C
Dichtheid: 1,276 g/cm³ (20 °C)
Vlampunt: 13°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000260
CAS nummer: 109-65-9
EINECS: 203-691-9
UN: 1126
ADR: 3,II
Merck Index: 14,01553
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90e7863ef88fa87359e8603e2390fade10d7abfd&vid=d84bd591b9efc2dde11d1f4430d85c98c591622d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR