1-Bromonaphthalene ≥95.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B0618-25G B0618-500G B0618-100G
TCIAB0618-25GEA 17.6 EUR
TCIAB0618-25G TCIAB0618-500G TCIAB0618-100G
1-Bromonaphthalene ≥95.0% (by GC) stabilised
1-Bromonaphthalene
Formule: C₁₀H₇Br
Kookpunt: 282°C
Smeltpunt: 0…6 °C
Dichtheid: 1,48 g/cm³ (20 °C)
Vlampunt: 66 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003868
CAS nummer: 90-11-9
EINECS: 201-965-2
Merck Index: 14,01425
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f643600463eb577931a1aa8e69f6360344d817c4&vid=f643600463eb577931a1aa8e69f6360344d817c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR