2-Bromopyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B0650-100ML B0650-25ML B0650-500ML
TCIAB0650-100MLEA 101 EUR
TCIAB0650-100ML TCIAB0650-25ML TCIAB0650-500ML
2-Bromopyridine ≥98.0% (by GC)
2-Bromopyridine
Formule: C₅H₄BrN
Kookpunt: 194°C
Dichtheid: 1,62 g/cm³ (20 °C)
Vlampunt: 54°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006219
CAS nummer: 109-04-6
EINECS: 203-641-6
UN: 2929
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4325252c307a8e2a91e4b2452010866a7fba0556&vid=4325252c307a8e2a91e4b2452010866a7fba0556&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR