Butyl methyl sulphide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B0729-5ML
TCIAB0729-5MLEA 124 EUR
TCIAB0729-5ML
Butyl methyl sulphide ≥98.0% (by GC)
Butyl methyl sulphide
Formule: C₅H₁₂S
MW: 104.21 g/mol
Kookpunt: 123°C
Vlampunt: 20°C
MDL Nummer: MFCD00015256
CAS nummer: 628-29-5
EINECS: 211-034-2
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR