Titanium(IV) butanolate

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B0742-25G B0742-500G
TCIAB0742-25GEA 18.7 EUR
TCIAB0742-25G TCIAB0742-500G
Titanium(IV) butanolate
Titanium(IV) butanolate
Formule: C₁₆H₃₆O₄Ti
Kookpunt: 310 °C (1013 hPa)
Smeltpunt: –55 °C
Dichtheid: 0,993 g/cm³ (20 °C)
Vlampunt: 46°C
MDL Nummer: MFCD00009433
CAS nummer: 5593-70-4
EINECS: 227-006-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51643979ff74e929fd24a23769e2908f61470110&vid=51643979ff74e929fd24a23769e2908f61470110&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Ignition residue 23.2 to 24.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR