Boterzuur ≥99.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Butanoic acid

B0754-25ML B0754-500ML
TCIAB0754-25MLEA 17.6 EUR
TCIAB0754-25ML TCIAB0754-500ML
Boterzuur ≥99.0% (by GC, titration analysis)
Boterzuur
Formule: C₄H₈O₂
MW: 88.11 g/mol
Kookpunt: 163°C
Smeltpunt: -3
Vlampunt: 75°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002814
CAS nummer: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: 8 III
Merck Index: 14,01593
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fb6bf5b2c2b02f030d161125b884712fad0b675&vid=1b2e897268a1c8e805190e03abd65cdc7d7c13c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR