Butyrylthiocholine iodide ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B0775-5G
TCIAB0775-5GEA 57.2 EUR
TCIAB0775-5G
Butyrylthiocholine iodide ≥99.0% (by titrimetric analysis)
Butyrylthiocholine iodide
Formule: H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I
Smeltpunt: 172 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00011845
CAS nummer: 1866-16-6
EINECS: 217-475-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce6b0f164944e4d07205274c354890b2e639fd99&vid=ce6b0f164944e4d07205274c354890b2e639fd99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 171.0 to 176.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR