2,1,3-Benzothiadiazole ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B0921-10G B0921-25G
TCIAB0921-10GEA 50.6 EUR
TCIAB0921-10G TCIAB0921-25G
2,1,3-Benzothiadiazole ≥99.0% (by GC)
2,1,3-Benzothiadiazole
Formule: C₆H₄N₂S
Kookpunt: 64°C
MDL Nummer: MFCD00005809
CAS nummer: 273-13-2
EINECS: 205-985-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=888a02729027d178c74578e4b02369ee846ca0a2&vid=888a02729027d178c74578e4b02369ee846ca0a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 43.0 to 47.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR