1,3-Bis(chloromethyl)tetramethyldisiloxane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B0978-25ML
TCIAB0978-25MLEA 81.4 EUR
TCIAB0978-25ML
1,3-Bis(chloromethyl)tetramethyldisiloxane ≥98.0% (by GC)
1,3-Bis(chloromethyl)tetramethyldisiloxane
Formule: C₆H₁₆Cl₂OSi₂
Kookpunt: 204°C
Vlampunt: 73°C
MDL Nummer: MFCD00000877
CAS nummer: 2362-10-9
EINECS: 219-109-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR