S-Butyl thiobenzoate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B1009-25ML
TCIAB1009-25MLEA 180 EUR
TCIAB1009-25ML
S-Butyl thiobenzoate ≥97.0% (by GC)
S-Butyl thiobenzoate
Formule: C₁₁H₁₄OS
Kookpunt: 160°C
MDL Nummer: MFCD00039955
CAS nummer: 7269-35-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR