1,2-Di(2-pyridyl)ethylene ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B1131-1G
TCIAB1131-1GEA 36.3 EUR
TCIAB1131-1G
1,2-Di(2-pyridyl)ethylene ≥98.0% (by GC, titration analysis)
1,2-Di(2-pyridyl)ethylene
Formule: C₁₂H₁₀N₂
Kookpunt: 347 °C (1013 hPa)
Smeltpunt: 119 °C
MDL Nummer: MFCD00006356
CAS nummer: 1437-15-6
EINECS: 215-866-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6e625f7d07bbd9036f213c2a690f2ecab44d6c5&vid=a6e625f7d07bbd9036f213c2a690f2ecab44d6c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 119.0 to 122.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR