Bis[(2S,3aR,4S,7aR)octahydro-7,8,8-trimethyl-4,7-methanobenzofuran-2-yl]ether ≥98.0% (by HPLC) for optical resolution

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (-)-MBF-OH Dimer, (-)-Noe's Reagent

B1220-100MG
TCIAB1220-100MGEA 49.5 EUR
TCIAB1220-100MG
Bis[(2S,3aR,4S,7aR)octahydro-7,8,8-trimethyl-4,7-methanobenzofuran-2-yl]ether ≥98.0% (by HPLC) for optical resolution
Bis[(2S,3aR,4S,7aR)octahydro-7,8,8-trimethyl-4,7-methanobenzofuran-2-yl]ether
Formule: C₂₄H₃₈O₃
MDL Nummer: MFCD00151131
CAS nummer: 108031-79-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 149.0 to 153.0 deg-C
Specific rotation [a]20/D -200.0 to -205.0 deg (C=2, THF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR