4-(4-Methylphenylthio)benzophenone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B1267-10G
TCIAB1267-10GEA 58.3 EUR
TCIAB1267-10G
4-(4-Methylphenylthio)benzophenone ≥98.0% (by GC)
4-(4-Methylphenylthio)benzophenone
Formule: C₂₀H₁₆OS
MDL Nummer: MFCD00055651
CAS nummer: 83846-85-9
EINECS: 281-064-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=626f5578165babf011e0c93914590bee0638da6c&vid=626f5578165babf011e0c93914590bee0638da6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 86.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR