4,4'-Biphenyldicarbonyl chloride ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B1308-10G
TCIAB1308-10GEA 163 EUR
TCIAB1308-10G
4,4'-Biphenyldicarbonyl chloride ≥97.0% (by GC, titration analysis)
4,4'-Biphenyldicarbonyl chloride
Formule: C₁₄H₈Cl₂O₂
MDL Nummer: MFCD00058934
CAS nummer: 2351-37-3
EINECS: 219-085-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystaline
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 186.0 to 190.0 deg-C
Solubility in hot Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR