2,2-Bis(4-chloroformyloxyphenyl)propane ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B1335-1G B1335-5G
TCIAB1335-1GEA 22 EUR
TCIAB1335-1G TCIAB1335-5G
2,2-Bis(4-chloroformyloxyphenyl)propane ≥97.0% (by GC, titration analysis)
2,2-Bis(4-chloroformyloxyphenyl)propane
Formule: C₁₇H₁₄Cl₂O₄
Kookpunt: 216°C
MDL Nummer: MFCD00060109
CAS nummer: 2024-88-6
EINECS: 217-970-8
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 93.0 to 96.0 deg-C
Solubility in Acetone almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR