Bisabolene mixture

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B1413-25ML
TCIAB1413-25MLEA 55 EUR
TCIAB1413-25ML
Bisabolene mixture
Bisabolene
Formule: C₁₅H₂₄
MDL Nummer: MFCD00129080
CAS nummer: 495-62-5

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) multi component mixture


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR