Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride monohydrate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B1626-25G B1626-5G
TCIAB1626-25GEA 57.2 EUR
TCIAB1626-25G TCIAB1626-5G
Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
Nalpha-Boc-L-arginine hydrochloride monohydrate
Formule: C₁₁H₂₂N₄O₄·HCl·H₂O
MDL Nummer: MFCD00063594
CAS nummer: 35897-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efab55fab4ee4e76d0b6c6d8ed2a3ed3e081e397&vid=94fdf9b1ff24f3a2e53774cb89a78ad14633bd2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0% (calcd.on anh.substance)
Specific rotation [a]20/D -7.0 to -9.0 deg (C=2, H2O)
Solubility in Water almost transparency
Water 4.5 to 6.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR