N-Benzylcinchonidinium chloride ≥98.0% (by HPLC, titration analysis) chiral phase-transfer catalyst

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B1683-10G
TCIAB1683-10GEA 134 EUR
TCIAB1683-10G
N-Benzylcinchonidinium chloride ≥98.0% (by HPLC, titration analysis) chiral phase-transfer catalyst
N-Benzylcinchonidinium chloride
Formule: C₂₆H₂₉ClN₂O
MDL Nummer: MFCD00082422
CAS nummer: 69257-04-1
EINECS: 273-938-3

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -182.0 to -188.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR