2-Butylaniline ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B1766-25G B1766-5G
TCIAB1766-25GEA 493 EUR
TCIAB1766-25G TCIAB1766-5G
2-Butylaniline ≥98.0% (by titrimetric analysis)
2-Butylaniline
Formule: C₁₀H₁₅N
Kookpunt: 126°C
MDL Nummer: MFCD00130033
CAS nummer: 2696-85-7
EINECS: 220-272-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64bee1670c5d8d513e2fabf15274f64b9b44637a&vid=64bee1670c5d8d513e2fabf15274f64b9b44637a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR