3-Bromo-p-anisaldehyde ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B1790-25G B1790-5G
TCIAB1790-25GEA 78.1 EUR
TCIAB1790-25G TCIAB1790-5G
3-Bromo-p-anisaldehyde ≥98.0% (by GC)
3-Bromo-p-anisaldehyde
Formule: C₈H₇BrO₂
Kookpunt: 162°C
Smeltpunt: 54 °C
MDL Nummer: MFCD00016599
CAS nummer: 34841-06-0
EINECS: 252-241-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a97862195eeff4a8ee4bbee455178619f3eeda7&vid=f7b3c8edc1b2418b434333131457034e5576e4b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR