2-Bromo-5-nitrotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B1853-25G B1853-5G
TCIAB1853-25GEA 245 EUR
TCIAB1853-25G TCIAB1853-5G
2-Bromo-5-nitrotoluene ≥98.0% (by GC)
2-Bromo-5-nitrotoluene
Formule: C₇H₆BrNO₂
Kookpunt: 143°C
Smeltpunt: 78 °C
MDL Nummer: MFCD00007281
CAS nummer: 7149-70-4
EINECS: 230-481-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=114528b994d9a94e06d61382d9123ef41b8e2898&vid=fff409429bbbd1d6efbed53371d99f4736ba8663&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR