2-Formylfenylboronzuur (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Formylbenzeneboronic acid, (2-Formylphenyl)metaboric acid, 2-Boronobenzaldehyde, (2-Formylphenyl)boronic acid, Dihydroxy(2-formylphenyl)borane

B1873-1G B1873-5G
TCIAB1873-1GEA 25.3 EUR
TCIAB1873-1G TCIAB1873-5G
2-Formylfenylboronzuur (contains varying amounts of Anhydride)
2-Formylfenylboronzuur
Formule: C₇H₇BO₃
MDL Nummer: MFCD00151822
CAS nummer: 40138-16-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=649ecf40736eb81e6741da4f00d3e69da2fae48e&vid=649ecf40736eb81e6741da4f00d3e69da2fae48e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (Neutralization titration) 97.0 to 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR